3,4-Dinitrobenzoic acid 98%

Supplier: Apollo Scientific
Warning

OR3469-25G OR3469-5G OR3469-1G
APOSOR3469-25GEA 358 EUR
APOSOR3469-25G APOSOR3469-5G APOSOR3469-1G
3,4-Dinitrobenzoic acid 98%
3,4-Dinitrobenzoic acid
Formula: C₇H₄N₂O₆
MW: 212,12 g/mol
Melting Pt: 163…167 °C
Storage Temperature: Ambient
MDL Number: MFCD00007255
CAS Number: 528-45-0
EINECS: 208-433-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR