2-Ethynyltoluene
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
1-Ethynyl-2-methylbenzene, 2-Methylphenylacetylene
OR3480-1G
OR3480-5G
OR3480-25G
APOSOR3480-1GEA
88.6
EUR
APOSOR3480-1G
APOSOR3480-5G
APOSOR3480-25G
2-Ethynyltoluene
2-Ethynyltoluene
Formula:
C₉H₈ MW: 116.1598 g/mol Boiling Pt: 50…60 °C/30mmHg Density: 0.922 g/cm³ (25 °C) Flash Pt: 35 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD03427202 CAS Number: 766-47-2 UN: 3295 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...