Benzyltriethylammonium bromide 98%

Supplier: Apollo Scientific
Warning

OR350020-100G OR350020-25G OR350020-500G
APOSOR350020-100GEA 94.5 EUR
APOSOR350020-100G APOSOR350020-25G APOSOR350020-500G
Benzyltriethylammonium bromide 98%
Benzyltriethylammonium bromide
Formula: C₁₃H₂₂BrN
MW: 272,23 g/mol
Melting Pt: 193…195 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00011822
CAS Number: 5197-95-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR