4-(Hydroxymethyl)benzyl alcohol 98%
Supplier: Apollo Scientific
Synonyms:
p-Phenylenedimethanol, 1,4-Benzenedimethanol, 1,4-Bis(hydroxymethyl)-benzene, p-Xylene-α,α'-diol
OR350022-500G
APOSOR350022-500GEA
78.3
EUR
APOSOR350022-500G
4-(Hydroxymethyl)benzyl alcohol 98%
4-(Hydroxymethyl)benzyl alcohol
Intermediate for organic synthesis; Synthesis of high performance resin.
Formula:
C₈H₁₀O₂ MW: 138.17 g/mol Boiling Pt: 138…143 °C/1mmHg Melting Pt: 114…118 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004665 CAS Number: 589-29-7 EINECS: 209-641-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...