Bicyclo[2.2.1]hept-5-ene-2,3-dicarboxylic acid 98%

Supplier: Apollo Scientific

Synonyms: 5-Norbornene-2,3-dicarboxylic acid

OR350049-5G OR350049-1G
APOSOR350049-5GEA 36.4 EUR
APOSOR350049-5G APOSOR350049-1G
Bicyclo[2.2.1]hept-5-ene-2,3-dicarboxylic acid 98%
Bicyclo[2.2.1]hept-5-ene-2,3-dicarboxylic acid
Formula: C₉H₁₀O₄
MW: 182,17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00152680
CAS Number: 3813-52-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR