3-Chloro-2-nitrotoluene
Supplier: Apollo Scientific
|
Danger
|
OR350086-25G
OR350086-5G
APOSOR350086-25GEA
111
EUR
APOSOR350086-25G
APOSOR350086-5G
3-Chloro-2-nitrotoluene
3-Chloro-2-nitrotoluene
Formula:
C₇H₆ClNO₂ MW: 171,58 g/mol Boiling Pt: 80 °C/2mmHg Density: 1,29 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD04117977 CAS Number: 5367-26-0 UN: 2433 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...