trans-2,5-Dimethylpiperazine 95%
Supplier: Apollo Scientific
OR350142-100G
APOSOR350142-100GEA
60.9
EUR
APOSOR350142-100G
trans-2,5-Dimethylpiperazine 95%
trans-2,5-Dimethylpiperazine
Formula:
C₆H₁₄N₂ MW: 114,19 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00070516 CAS Number: 2815-34-1 UN: 2926 ADR: 4.1+6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...