N-ɑ-Fmoc-N-ε-boc-L-lysine 97%

Supplier: Apollo Scientific
Warning

Synonyms: N-ε-(tert-Butoxycarbonyl)-N-ɑ-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-lysine, (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-6-((tert-butoxycarbonyl)amino)hexanoic acid, N-ε-Boc-N-ɑ-Fmoc-L-lysine, Fmoc-Lys(Boc)-OH

OR350184-100G OR350184-25G OR350184-500G
APOSOR350184-100GEA 75.4 EUR
APOSOR350184-100G APOSOR350184-25G APOSOR350184-500G
N-ɑ-Fmoc-N-ε-boc-L-lysine 97%
N-alpha-Fmoc-N-epsilon-boc-L-lysine
Formula: C₂₆H₃₂N₂O₆
MW: 468.55 g/mol
Melting Pt: 129…135 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00037138
CAS Number: 71989-26-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR