2-(Methylthio)phenol 98%
Supplier: Apollo Scientific
Synonyms:
2-Hydroxythioanisole
OR350271-25G
APOSOR350271-25GEA
120
EUR
APOSOR350271-25G
2-(Methylthio)phenol 98%
2-(Methylthio)phenol
Formula:
C₇H₈OS MW: 140,21 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00002211 CAS Number: 1073-29-6 |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...