2-Methoxythiazole

Supplier: Apollo Scientific

OR350279-25G OR350279-100G
APOSOR350279-25GEA 37.7 EUR
APOSOR350279-25G APOSOR350279-100G
2-Methoxythiazole
2-Methoxythiazole
Formula: C₄H₅NOS
MW: 115,16 g/mol
Boiling Pt: 150…151 °C
Density: 1,2 g/cm³ (25 °C)
Flash Pt: 49 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01631143
CAS Number: 14542-13-3
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR