1-N-Boc-4-(4-bromophenyl)piperidine

Supplier: Apollo Scientific

Synonyms: 1-N-Boc-4-(4-Bromophenyl)piperidine

OR350304-5G
APOSOR350304-5GEA 105 EUR
APOSOR350304-5G
1-N-Boc-4-(4-bromophenyl)piperidine
1-N-Boc-4-(4-bromophenyl)piperidine
Formula: C₁₆H₂₂BrNO₂
MW: 340,2600 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09878774
CAS Number: 769944-78-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR