2-Propoxybenzoic acid

Supplier: Apollo Scientific

OR350317-100G OR350317-25G OR350317-500G
APOSOR350317-100GEA 91.6 EUR
APOSOR350317-100G APOSOR350317-25G APOSOR350317-500G
2-Propoxybenzoic acid
2-Propoxybenzoic acid
Formula: C₁₀H₁₂O₃
Storage Temperature: Ambient
MDL Number: MFCD01075687
CAS Number: 2100-31-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR