3,5,6-Trichlorosalicylic acid 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
3,5,6-Trichlorosalicylic Acid
OR350376-100G
OR350376-25G
APOSOR350376-100GEA
48
EUR
APOSOR350376-100G
APOSOR350376-25G
3,5,6-Trichlorosalicylic acid 97%
3,5,6-Trichlorosalicylic acid
Formula:
C₇H₃Cl₃O₃ MW: 241,46 g/mol Melting Pt: 209…211 °C Storage Temperature: Ambient |
MDL Number:
MFCD00075245 CAS Number: 40932-60-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...