Ethyl-5-methyl-1H-pyrrole-2-carboxylate 97+%
Supplier: Apollo Scientific
Synonyms:
Ethyl 5-methyl-1H-pyrrole-2-carboxylate
OR350470-25G
OR350470-1G
APOSOR350470-25GEA
201
EUR
APOSOR350470-25G
APOSOR350470-1G
Ethyl-5-methyl-1H-pyrrole-2-carboxylate 97+%
Ethyl-5-methyl-1H-pyrrole-2-carboxylate
Formula:
C₈H₁₁NO₂ MW: 153,1800 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01006731 CAS Number: 3284-51-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...