4-((3-Hydroxypropyl)amino)-3-nitrophenol 95+%
Supplier: Apollo Scientific
OR350491-100G
OR350491-25G
OR350491-5G
APOSOR350491-100GEA
109
EUR
APOSOR350491-100G
APOSOR350491-25G
APOSOR350491-5G
4-((3-Hydroxypropyl)amino)-3-nitrophenol 95+%
4-((3-Hydroxypropyl)amino)-3-nitrophenol
Formula:
C₉H₁₂N₂O₄ MW: 212,2000 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07368637 CAS Number: 92952-81-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...