Methyl-3-amino-4-bromobenzoate 98%

Supplier: Apollo Scientific

Synonyms: Methyl 3-amino-4-bromobenzoate

OR350503-100G OR350503-25G
APOSOR350503-100GEA 238 EUR
APOSOR350503-100G APOSOR350503-25G
Methyl-3-amino-4-bromobenzoate 98%
Methyl 3-amino-4-bromobenzoate
Formula: C₈H₈BrNO₂
MW: 230,0586 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00553090
CAS Number: 46064-79-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR