3-Bromo-6-chloropicolinic acid 97%

Supplier: Apollo Scientific
Warning

Synonyms: 3-Bromo-6-chloropyridine-2-carboxylic acid

OR3520-1G OR3520-5G OR3520-10G
APOSOR3520-1GEA 21.9 EUR
APOSOR3520-1G APOSOR3520-5G APOSOR3520-10G
3-Bromo-6-chloropicolinic acid 97%
3-Bromo-6-chloropicolinic acid
Formula: C₆H₃BrClNO₂
MW: 236.4505 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09258777
CAS Number: 929000-66-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR