2-(3-Bromophenyl)ethanol

Supplier: Apollo Scientific
Warning

OR3522-1G OR3522-5G
APOSOR3522-1GEA 49.5 EUR
APOSOR3522-1G APOSOR3522-5G
2-(3-Bromophenyl)ethanol
2-(3-Bromophenyl)ethanol
Formula: C₈H₉BrO
MW: 201,06 g/mol
Boiling Pt: 107…110 °C/1mmHg
Density: 1,478 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00191857
CAS Number: 28229-69-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR