5-Bromo-2-hydroxy-6-picoline 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
3-Bromo-6-hydroxy-2-methylpyridine, 5-Bromo-6-methylpyridin-2-ol
OR3528-1G
OR3528-25G
OR3528-5G
APOSOR3528-1GEA
21.9
EUR
APOSOR3528-1G
APOSOR3528-25G
APOSOR3528-5G
5-Bromo-2-hydroxy-6-picoline 98%
5-Bromo-2-hydroxy-6-picoline
Formula:
C₆H₆BrNO MW: 188,02 g/mol Melting Pt: 199…203 °C Storage Temperature: Ambient |
MDL Number:
MFCD03427653 CAS Number: 54923-31-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...