7-Aminoindole 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Indol-7-ylamine, 1H-Indol-7-amine, 7-Amino-1H-indole, 7-Indolamine
OR3531-100MG
OR3531-500MG
APOSOR3531-100MGEA
24.8
EUR
APOSOR3531-100MG
APOSOR3531-500MG
7-Aminoindole 97%
7-Aminoindole
Formula:
C₈H₈N₂ MW: 132,16 g/mol Melting Pt: 90…100 °C Storage Temperature: Freezer |
MDL Number:
MFCD02093960 CAS Number: 5192-04-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...