Isoquinoline-8-amine 98%

Supplier: Apollo Scientific
Warning

Synonyms: Isoquinolin-8-amine, 8-Aminoisoquinoline

OR3549-1G OR3549-5G
APOSOR3549-1GEA 49.5 EUR
APOSOR3549-1G APOSOR3549-5G
Isoquinoline-8-amine 98%
Isoquinoline-8-amine
Formula: C₉H₈N₂
MW: 144,1700 g/mol
Melting Pt: 168…169 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00179553
CAS Number: 23687-27-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR