3-Propoxyphenylboronic acid 98%

Supplier: Apollo Scientific

OR360035-5G
APOSOR360035-5GEA 66.7 EUR
APOSOR360035-5G
3-Propoxyphenylboronic acid 98%
3-Propoxyphenylboronic acid
Formula: C₉H₁₃BO₃
MW: 180,0100 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03427190
CAS Number: 149557-18-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR