4-Chloro-3-(cyclopropylcarbamoyl)phenylboronic acid 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
4-Chloro-3-(cyclopropylcarbamoyl)benzeneboronic acid, (4-Chloro-3-(cyclopropylcarbamoyl)phenyl)boronic acid
OR360076-1G
OR360076-5G
APOSOR360076-1GEA
285
EUR
APOSOR360076-1G
APOSOR360076-5G
4-Chloro-3-(cyclopropylcarbamoyl)phenylboronic acid 98%
4-Chloro-3-(cyclopropylcarbamoyl)phenylboronic acid
Formula:
C₁₀H₁₁BClNO₃ MW: 239,46 g/mol Melting Pt: 238…242 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD07783865 CAS Number: 871332-73-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...