6-Indoleboronic acid pinacol ester 95+%

Supplier: Apollo Scientific

Synonyms: 6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole

OR361278-5G
APOSOR361278-5GEA 207 EUR
APOSOR361278-5G
6-Indoleboronic acid pinacol ester 95+%
6-Indoleboronic acid pinacol ester
Formula: C₁₄H₁₈BNO₂
MW: 243,1092 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07368852
CAS Number: 642494-36-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR