4-Amino-6-methoxy-2-methylquinoline
Supplier: Apollo Scientific
|
Warning
|
OR3658-1G
OR3658-5G
APOSOR3658-1GEA
335
EUR
APOSOR3658-1G
APOSOR3658-5G
4-Amino-6-methoxy-2-methylquinoline
4-Amino-6-methoxy-2-methylquinoline
Formula:
C₁₁H₁₂N₂O Melting Pt: 202…203 °C Storage Temperature: Ambient |
MDL Number:
MFCD00552889 CAS Number: 104217-23-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...