6-Bromo-1H-indazole 98%
Supplier: Apollo Scientific
|
Danger
|
OR3676-25G
OR3676-5G
APOSOR3676-25GEA
49.5
EUR
APOSOR3676-25G
APOSOR3676-5G
6-Bromo-1H-indazole 98%
6-Bromo-1H-indazole
Formula:
C₇H₅BrN₂ MW: 197,0300 g/mol Melting Pt: 180…186 °C Storage Temperature: Ambient |
MDL Number:
MFCD03265457 CAS Number: 79762-54-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...