4-Chloro-1H-indazole
Supplier: Apollo Scientific
|
Danger
|
OR3679-1G
OR3679-250MG
OR3679-5G
APOSOR3679-1GEA
16.1
EUR
APOSOR3679-1G
APOSOR3679-250MG
APOSOR3679-5G
4-Chloro-1H-indazole
4-Chloro-1H-indazole
Formula:
C₇H₅ClN₂ MW: 152,58 g/mol Melting Pt: 155…160 °C Storage Temperature: Ambient |
MDL Number:
MFCD00211065 CAS Number: 13096-96-3 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...