3-Chloro-4-methylbenzonitrile 98%

Supplier: Apollo Scientific
Danger

OR3687-25G OR3687-5G OR3687-100G
APOSOR3687-25GEA 34.8 EUR
APOSOR3687-25G APOSOR3687-5G APOSOR3687-100G
3-Chloro-4-methylbenzonitrile 98%
3-Chloro-4-methylbenzonitrile
Formula: C₈H₆ClN
MW: 151,60 g/mol
Melting Pt: 46…48 °C
Flash Pt: 84 °C
Storage Temperature: Ambient
MDL Number: MFCD00001799
CAS Number: 21423-81-4
EINECS: 244-381-3
UN: 1325
ADR: 4.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR