4-Methyl-1H-indazol-7-amine
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
7-Amino-4-methyl-1H-indazole
OR3691-1G
OR3691-5G
OR3691-250MG
APOSOR3691-1GEA
59.6
EUR
APOSOR3691-1G
APOSOR3691-5G
APOSOR3691-250MG
4-Methyl-1H-indazol-7-amine
4-Methyl-1H-indazol-7-amine
Formula:
C₈H₉N₃ MW: 147,18 g/mol Melting Pt: 211…214 °C Storage Temperature: Ambient |
MDL Number:
MFCD09607935 CAS Number: 952183-44-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...