4'-Bromobiphenyl-4-ylboronic acid
Supplier: Apollo Scientific
Synonyms:
4'-Bromo-4-biphenylboronic acid
OR3708-1G
OR3708-5G
APOSOR3708-1GEA
69.8
EUR
APOSOR3708-1G
APOSOR3708-5G
4'-Bromobiphenyl-4-ylboronic acid
4'-Bromobiphenyl-4-ylboronic acid
Formula:
C₁₂H₁₀BBrO₂ MW: 276,9216 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02251652 CAS Number: 480996-05-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...