2-Chloro-7H-purin-6-amine
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
6-Amino-2-chloropurine, 2-Chloroadenine
OR3724-1G
OR3724-5G
OR3724-10G
APOSOR3724-1GEA
21.9
EUR
APOSOR3724-1G
APOSOR3724-5G
APOSOR3724-10G
2-Chloro-7H-purin-6-amine
2-Chloro-7H-purin-6-amine
Formula:
C₅H₄ClN₅ Melting Pt: 384…389 °C Storage Temperature: Ambient |
MDL Number:
MFCD00037925 CAS Number: 1839-18-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...