3-Amino-4-methylphenylboronic acid

Supplier: Apollo Scientific
Warning

Synonyms: 3-Amino-4-methylbenzeneboronic Acid, 3-Amino-p-tolylboronic Acid, 3-Amino-4-methylphenylboronic Acid

OR3743-5G OR3743-1G
APOSOR3743-5GEA 74.2 EUR
APOSOR3743-5G APOSOR3743-1G
3-Amino-4-methylphenylboronic acid
3-Amino-4-methylphenylboronic acid
Formula: C₇H₁₀BNO₂
MW: 150,97 g/mol
Melting Pt: 250 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01074640
CAS Number: 22237-12-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR