α,4-Dichlorotoluene 99%
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
1-Chloro-4-(chloromethyl)benzene, 4-Chlorobenzyl chloride
OR3764-1KG
OR3764-250G
OR3764-5KG
OR3764-25G
OR3764-100G
OR3764-400G
APOSOR3764-1KGEA
104
EUR
APOSOR3764-1KG
APOSOR3764-250G
APOSOR3764-5KG
APOSOR3764-25G
APOSOR3764-100G
APOSOR3764-400G
α,4-Dichlorotoluene 99%
α,4-Dichlorotoluene
Formula:
C₇H₆Cl₂ MW: 161,03 g/mol Boiling Pt: 216…222 °C Melting Pt: 26…30 °C Flash Pt: 107 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000914 CAS Number: 104-83-6 EINECS: 203-242-7 UN: 3427 ADR: 6.1,III |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...