4-(2,3-Dimethylphenylcarbamoyl)phenylboronic acid 98%

Supplier: Apollo Scientific
Warning

OR3792-1G OR3792-5G OR3792-250MG
APOSOR3792-1GEA 94.4 EUR
APOSOR3792-1G APOSOR3792-5G APOSOR3792-250MG
4-(2,3-Dimethylphenylcarbamoyl)phenylboronic acid 98%
4-(2,3-Dimethylphenylcarbamoyl)phenylboronic acid
Formula: C₁₅H₁₆BNO₃
Storage Temperature: Refrigerator
MDL Number: MFCD09027210
CAS Number: 913835-36-6

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR