5-Hydroxy-2-methoxylpyridine 98%
Supplier: Apollo Scientific
|
Warning
|
OR3808-10G
OR3808-5G
OR3808-1G
APOSOR3808-10GEA
137
EUR
APOSOR3808-10G
APOSOR3808-5G
APOSOR3808-1G
5-Hydroxy-2-methoxylpyridine 98%
5-Hydroxy-2-methoxylpyridine
Formula:
C6H7NO2 MW: 125,1300 g/mol Melting Pt: 80…81 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD06858737 CAS Number: 51834-97-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...