2,4-Dichloroquinoline 98%

Supplier: Apollo Scientific
Danger

OR3822-1G OR3822-25G OR3822-5G
APOSOR3822-1GEA 21.9 EUR
APOSOR3822-1G APOSOR3822-25G APOSOR3822-5G
2,4-Dichloroquinoline 98%
2,4-Dichloroquinoline
Formula: C₉H₅Cl₂N
MW: 198,0500 g/mol
Melting Pt: 62…67 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00023939
CAS Number: 703-61-7
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR