6-Nitrophthalide 97%

Supplier: Apollo Scientific
Warning

OR3915-100G OR3915-25G OR3915-5G
APOSOR3915-100GEA 256 EUR
APOSOR3915-100G APOSOR3915-25G APOSOR3915-5G
6-Nitrophthalide 97%
6-Nitrophthalide
Formula: C₈H₅NO₄
Melting Pt: 140…145 °C
Storage Temperature: Ambient
MDL Number: MFCD00033529
CAS Number: 610-93-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR