3-(1-Pyrrolidinylcarbonyl)phenylboronic acid 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
(3-(Pyrrolidine-1-carbonyl)phenyl)boronic acid
OR3976-1G
OR3976-5G
APOSOR3976-1GEA
85.8
EUR
APOSOR3976-1G
APOSOR3976-5G
3-(1-Pyrrolidinylcarbonyl)phenylboronic acid 97%
3-(1-Pyrrolidinylcarbonyl)phenylboronic acid
Formula:
C₁₁H₁₄BNO₃ MW: 219,0500 g/mol Melting Pt: 113…116 °C Storage Temperature: Ambient |
MDL Number:
MFCD04115685 CAS Number: 723281-53-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...