3-Bromo-5-chlorophenol 97%

Supplier: Apollo Scientific
Warning

OR399004-10G OR399004-25G
APOSOR399004-10GEA 87.3 EUR
APOSOR399004-10G APOSOR399004-25G
3-Bromo-5-chlorophenol 97%
3-Bromo-5-chlorophenol
Formula: C₆H₄BrClO
MW: 207,45 g/mol
Melting Pt: 66…70 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07780676
CAS Number: 56962-04-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR