2-Bromo-5-methoxyphenol 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
4-Brom-3-hydroxyanisole
OR399007-10G
OR399007-5G
APOSOR399007-10GEA
104
EUR
APOSOR399007-10G
APOSOR399007-5G
2-Bromo-5-methoxyphenol 98%
2-Bromo-5-methoxyphenol
Formula:
C₇H₇BrO₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD03428523 CAS Number: 63604-94-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...