3-Bromo-4-methoxyphenol 98%

Supplier: Apollo Scientific
Warning

OR399010-25G
APOSOR399010-25GEA 185 EUR
APOSOR399010-25G
3-Bromo-4-methoxyphenol 98%
3-Bromo-4-methoxyphenol
Formula: C7H7BrO2
MW: 203,0300 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11054143
CAS Number: 17332-12-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR