2,6-Dichloro-3-nitrobenzoic acid 98%

Supplier: Apollo Scientific

OR39999-5G
APOSOR39999-5GEA 58 EUR
APOSOR39999-5G
2,6-Dichloro-3-nitrobenzoic acid 98%
2,6-Dichloro-3-nitrobenzoic acid
Formula: C₇H₃Cl₂NO₄
MW: 236,0100 g/mol
Melting Pt: 152…158 °C
Storage Temperature: Ambient
MDL Number: MFCD00075422
CAS Number: 55775-97-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR