4-Bromo-2-iodotoluene 98%

Supplier: Apollo Scientific
Warning

OR400110-25G OR400110-5G
APOSOR400110-25GEA 33 EUR
APOSOR400110-25G APOSOR400110-5G
4-Bromo-2-iodotoluene 98%
4-Bromo-2-iodotoluene
Formula: C₇H₆BrI
Storage Temperature: Ambient
MDL Number: MFCD07780671
CAS Number: 260558-15-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR