4-Iodo-3-nitrobenzoic acid 97%
Supplier: Apollo Scientific
OR400150-1G
OR400150-5G
OR400150-25G
APOSOR400150-1GEA
24.8
EUR
APOSOR400150-1G
APOSOR400150-5G
APOSOR400150-25G
4-Iodo-3-nitrobenzoic acid 97%
4-Iodo-3-nitrobenzoic acid
Formula:
C₇H₄INO₄ MW: 293,02 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00502399 CAS Number: 35674-27-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...