2-Amino-3-chlorophenol
Supplier: Apollo Scientific
|
Warning
|
OR400160-5G
APOSOR400160-5GEA
186
EUR
APOSOR400160-5G
2-Amino-3-chlorophenol
2-Amino-3-chlorophenol
Formula:
C₆H₆ClNO MW: 143,5700 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11848418 CAS Number: 56962-00-6 UN: 2811 ADR: 6.1,III |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...