6-Bromoisobenzofuran-1(3H)-one

Supplier: Apollo Scientific

Synonyms: 6-Bromophthalide

OR400356-5G
APOSOR400356-5GEA 85.7 EUR
APOSOR400356-5G
6-Bromoisobenzofuran-1(3H)-one
6-Bromoisobenzofuran-1(3H)-one
Formula: C₈H₅BrO₂
MW: 213,0281 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03428561
CAS Number: 19477-73-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR