2-Amino-5-bromobenzylalkohol

Supplier: Apollo Scientific

Synonyms: (2-Amino-5-bromophenyl)methanol

OR400402-1G OR400402-5G
APOSOR400402-1GEA 61.1 EUR
APOSOR400402-1G APOSOR400402-5G
2-Amino-5-bromobenzylalkohol
2-Amino-5-bromobenzylalkohol
Formula: C₇H₈BrNO
MW: 202,0500 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08234509
CAS Number: 20712-12-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR