3-Iodo-4-nitrophenol

Supplier: Apollo Scientific

OR400409-1G OR400409-5G
APOSOR400409-1GEA 94.5 EUR
APOSOR400409-1G APOSOR400409-5G
3-Iodo-4-nitrophenol
3-Iodo-4-nitrophenol
Formula: C₆H₄INO₃
MW: 265,01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11617270
CAS Number: 50590-07-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR