2-Hydroxypyrazine 95%

Supplier: Apollo Scientific
Warning

Synonyms: Pyrazin-2(1H)-one, 2(1H)-Pyrazinone, 2-Pyrazinol

OR40046-1G OR40046-5G
APOSOR40046-1GEA 14.7 EUR
APOSOR40046-1G APOSOR40046-5G
2-Hydroxypyrazine 95%
2-Hydroxypyrazine
Formula: C₄H₄N₂O
MW: 96,08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00233966
CAS Number: 6270-63-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR