2-Bromo-3-nitrobenzaldehyde

Supplier: Apollo Scientific
Warning

OR400521-1G OR400521-5G OR400521-10G
APOSOR400521-1GEA 37.7 EUR
APOSOR400521-1G APOSOR400521-5G APOSOR400521-10G
2-Bromo-3-nitrobenzaldehyde
2-Bromo-3-nitrobenzaldehyde
Formula: C₇H₄BrNO₃
Storage Temperature: Ambient
MDL Number: MFCD15527595
CAS Number: 90407-21-9
UN: 3077
ADR: 9+EHS,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR